Learn More
Medchemexpress LLC Danofloxacin mesylate | 119478-55-6 | 99.88% | 1 G
SDP
Catalog No. 5000405274
Encompass_Preferred
Danofloxacin mesylate (CP 76136-27) is a fluoroquinolone veterinary drug with broad-spectrum bactericidal activity. It primarily inhibits the DNA gyrase of bacteria and is a substrate for ATP-dependent efflux transporters (P-gp and MRP2). It is designed for laboratory research.
  • Fluoroquinolone veterinary drug
  • Broad-spectrum bactericidal activity
  • Inhibits bacterial DNA gyrase
  • Substrate for ATP-dependent efflux transporters (P-gp and MRP2)
Catalog No. 50-004-05274 Supplier Medchemexpress LLC Supplier No. HYB05011G
Danofloxacin mesylate (CP 76136-27) is a fluoroquinolone veterinary drug with broad-spectrum bactericidal activity. It primarily inhibits the DNA gyrase of bacteria and is a substrate for ATP-dependent efflux transporters (P-gp and MRP2). It is designed for laboratory research.
  • Fluoroquinolone veterinary drug
  • Broad-spectrum bactericidal activity
  • Inhibits bacterial DNA gyrase
  • Substrate for ATP-dependent efflux transporters (P-gp and MRP2)