Learn More
Medchemexpress LLC Danofloxacin mesylate | 119478-55-6 | 99.9% | 25 MG
SDP
Catalog No. 5000450548
Encompass_Preferred
Danofloxacin mesylate is a fluoroquinolone veterinary drug known for its broad-spectrum bactericidal activity. It primarily functions by inhibiting the DNA gyrase of bacteria and also acts as a substrate for ATP-dependent efflux transporters such as P-gp and MRP2. This compound is intended for research use only.
  • Fluoroquinolone veterinary drug
  • Broad-spectrum bactericidal activity
  • Inhibits bacterial DNA gyrase
  • Substrate for ATP-dependent efflux transporters (P-gp and MRP2)
  • Exhibits asymmetric transport across Caco-2 cells
  • Inhibits bovine bacterial respiratory pathogens
Catalog No. 50-004-50548 Supplier Medchemexpress LLC Supplier No. 25MGHYB0501
Danofloxacin mesylate is a fluoroquinolone veterinary drug known for its broad-spectrum bactericidal activity. It primarily functions by inhibiting the DNA gyrase of bacteria and also acts as a substrate for ATP-dependent efflux transporters such as P-gp and MRP2. This compound is intended for research use only.
  • Fluoroquinolone veterinary drug
  • Broad-spectrum bactericidal activity
  • Inhibits bacterial DNA gyrase
  • Substrate for ATP-dependent efflux transporters (P-gp and MRP2)
  • Exhibits asymmetric transport across Caco-2 cells
  • Inhibits bovine bacterial respiratory pathogens