Learn More
Medchemexpress LLC Danofloxacin mesylate | 119478-55-6 | 99.9% | 50 MG
SDP
Catalog No. 5000405271
Encompass_Preferred
Danofloxacin mesylate is a fluoroquinolone used as a veterinary drug. It exhibits a broad spectrum of bactericidal activity, primarily by inhibiting the DNA cycloenzyme of bacteria. Additionally, it acts as a substrate for ATP-dependent efflux transporters (P-gp and MRP2).
  • Used as a veterinary drug
  • Exhibits a broad spectrum of bactericidal activity
  • Primarily inhibits bacterial DNA cycloenzyme
  • Acts as a substrate for ATP-dependent efflux transporters (P-gp and MRP2)
Catalog No. 50-004-05271 Supplier Medchemexpress LLC Supplier No. HYB050150MG
Danofloxacin mesylate is a fluoroquinolone used as a veterinary drug. It exhibits a broad spectrum of bactericidal activity, primarily by inhibiting the DNA cycloenzyme of bacteria. Additionally, it acts as a substrate for ATP-dependent efflux transporters (P-gp and MRP2).
  • Used as a veterinary drug
  • Exhibits a broad spectrum of bactericidal activity
  • Primarily inhibits bacterial DNA cycloenzyme
  • Acts as a substrate for ATP-dependent efflux transporters (P-gp and MRP2)