Learn More
Medchemexpress LLC Danofloxacin mesylate | 119478-55-6 | 99.9% | 500 MG
SDP
Catalog No. 5000409289
Encompass_Preferred
Danofloxacin mesylate is a fluoroquinolone veterinary drug with a broad spectrum of bactericidal activity. Its primary mechanism of action involves inhibiting bacterial DNA gyrase. Additionally, it acts as a substrate for ATP-dependent efflux transporters, specifically P-gp and MRP2.
  • Fluoroquinolone veterinary drug
  • Broad spectrum bactericidal activity
  • Inhibits bacterial DNA gyrase
  • Substrate for ATP-dependent efflux transporters (P-gp and MRP2)
  • Intended for research use only
  • Inhibits bovine bacterial respiratory pathogens (e.g., Pasteurella hemolyticus, Pasteurella multocida, and Haemophilus somnosus)
  • Exhibits asymmetric transport across Caco-2 cells
Catalog No. 50-004-09289 Supplier Medchemexpress LLC Supplier No. HYB0501500MG
Danofloxacin mesylate is a fluoroquinolone veterinary drug with a broad spectrum of bactericidal activity. Its primary mechanism of action involves inhibiting bacterial DNA gyrase. Additionally, it acts as a substrate for ATP-dependent efflux transporters, specifically P-gp and MRP2.
  • Fluoroquinolone veterinary drug
  • Broad spectrum bactericidal activity
  • Inhibits bacterial DNA gyrase
  • Substrate for ATP-dependent efflux transporters (P-gp and MRP2)
  • Intended for research use only
  • Inhibits bovine bacterial respiratory pathogens (e.g., Pasteurella hemolyticus, Pasteurella multocida, and Haemophilus somnosus)
  • Exhibits asymmetric transport across Caco-2 cells