Learn More
Medchemexpress LLC Danofloxacin mesylate | 119478-55-6 | MFCD00673687 | 99.9% | 100 MG
SDP
Catalog No. 5000409288
Encompass_Preferred
Danofloxacin mesylate (CP 76136-27) is a fluoroquinolone used as a veterinary drug. It exhibits broad-spectrum bactericidal activity primarily by inhibiting bacterial DNA gyrase and also functions as a substrate for ATP-dependent efflux transporters such as P-gp and MRP2.
  • Used as a veterinary drug
  • Broad-spectrum bactericidal activity
  • Inhibits bacterial DNA gyrase
  • Substrate for ATP-dependent efflux transporters (P-gp and MRP2)
  • Targets P-glycoprotein and bacterial components
  • Involves membrane transporter/ion channel and anti-infection mechanisms
  • Effective against bovine bacterial respiratory pathogens
  • Demonstrates asymmetric transport across Caco-2 cells
Catalog No. 50-004-09288 Supplier Medchemexpress LLC Supplier No. HYB0501100MG
Danofloxacin mesylate (CP 76136-27) is a fluoroquinolone used as a veterinary drug. It exhibits broad-spectrum bactericidal activity primarily by inhibiting bacterial DNA gyrase and also functions as a substrate for ATP-dependent efflux transporters such as P-gp and MRP2.
  • Used as a veterinary drug
  • Broad-spectrum bactericidal activity
  • Inhibits bacterial DNA gyrase
  • Substrate for ATP-dependent efflux transporters (P-gp and MRP2)
  • Targets P-glycoprotein and bacterial components
  • Involves membrane transporter/ion channel and anti-infection mechanisms
  • Effective against bovine bacterial respiratory pathogens
  • Demonstrates asymmetric transport across Caco-2 cells