Learn More
TARGETMOL CHEMICALS INC Danofloxacin mesylate 500MG
SDP
Catalog No. 502878323
Encompass_Preferred
Also available in 1 g, 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Danofloxacin mesylate (CP 76136-27) is a synthetic antibacterial agent of the fluoroquinolone class that primarily inhibits bacterial DNA-gyrase. Purity 99.66%
Catalog No. 50-287-8323 Supplier TARGETMOL CHEMICALS INC Supplier No. T1276500MG
Also available in 1 g, 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Danofloxacin mesylate (CP 76136-27) is a synthetic antibacterial agent of the fluoroquinolone class that primarily inhibits bacterial DNA-gyrase. Purity 99.66%