Learn More
TARGETMOL CHEMICALS INC DANOPREVIR 5MG
SDP
Catalog No. 502576624
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Danoprevir (RG7227) is a peptidomimetic inhibitor of the NS3/4A protease(IC50=0.2-3.5 nM) of hepatitis C virus (HCV). purity: 99%
Catalog No. 50-257-6624 Supplier TARGETMOL CHEMICALS INC Supplier No. T60255MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Danoprevir (RG7227) is a peptidomimetic inhibitor of the NS3/4A protease(IC50=0.2-3.5 nM) of hepatitis C virus (HCV). purity: 99%