Learn More
Selleck Chemical LLC Danoprevir ITMN-191-2mg
SDP
Supplier:  Selleck Chemical LLC S11832MG
Encompass_Preferred
Danoprevir (ITMN-191, RG7227) is a peptidomimetic inhibitor of the NS3/4A protease of hepatitis C virus (HCV) with IC50 of 0.2-3.5 nM, inhibition effect for HCV genotypes 1A/1B/4/5/6 is ~10-fold higher than 2B/3A. Phase 2.
Catalog No. 50-238-9785