Learn More
Apexbio Technology LLC Danoprevir (RG7227), 5mg. Cas: 850876-88-9 MFCD: MFCD16038038. HCV NS3/4A protease inhibitor
SDP
Catalog No. 501904862
Encompass_Preferred
Danoprevir (R7227) is a potent and selective inhibitor of Hepatitis C Virus (HCV) NS3/4A protease, a chymotrypsin-like serine protease playing an essential role in the viral replication process of HCV, that non-covalently binds to and hence inhibits HCV NS3 protease. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4862 Supplier Apexbio Technology LLC Supplier No. A40245
Danoprevir (R7227) is a potent and selective inhibitor of Hepatitis C Virus (HCV) NS3/4A protease, a chymotrypsin-like serine protease playing an essential role in the viral replication process of HCV, that non-covalently binds to and hence inhibits HCV NS3 protease. Other sizes are available. Please inqury us for quote.