Learn More
Medchemexpress LLC DANP dihydrochloride | 761401-89-2 | 99.3% | 100 MG
SDP
Catalog No. 5000341293
Encompass_Preferred
DANP dihydrochloride is a bulge base recognition probe that strongly and specifically binds to single cytosine and thymine bulges.
  • Functions as a bulge base recognition probe
  • Strongly and specifically binds to single cytosine and thymine bulges
  • Can stabilize both single cytosine and thymine bulges in duplex DNA
Catalog No. 50-003-41293 Supplier Medchemexpress LLC Supplier No. HY129594A100MG
DANP dihydrochloride is a bulge base recognition probe that strongly and specifically binds to single cytosine and thymine bulges.
  • Functions as a bulge base recognition probe
  • Strongly and specifically binds to single cytosine and thymine bulges
  • Can stabilize both single cytosine and thymine bulges in duplex DNA