Learn More
Medchemexpress LLC DANP dihydrochloride | 761401-89-2 | 99.3% | 25 MG
SDP
Catalog No. 5000339194
Encompass_Preferred
DANP dihydrochloride is a bulge base recognition probe that strongly and specifically binds to single cytosine and thymine bulges. It can stabilize both single cytosine and thymine bulges in duplex DNA.
  • Bulge base recognition probe
  • Strongly and specifically binds to single cytosine and thymine bulges
  • Stabilizes both single cytosine and thymine bulges in duplex DNA
  • Fluorescent dye
Catalog No. 50-003-39194 Supplier Medchemexpress LLC Supplier No. 25MGHY129594A
DANP dihydrochloride is a bulge base recognition probe that strongly and specifically binds to single cytosine and thymine bulges. It can stabilize both single cytosine and thymine bulges in duplex DNA.
  • Bulge base recognition probe
  • Strongly and specifically binds to single cytosine and thymine bulges
  • Stabilizes both single cytosine and thymine bulges in duplex DNA
  • Fluorescent dye