Learn More
Medchemexpress LLC Danp dihydrochloride | 99.3% | 10 MG
SDP
Catalog No. 5000339978
Encompass_Preferred
DANP dihydrochloride is a bulge base recognition probe that strongly and specifically binds to the single cytosine and thymine bulges. It can stabilize both single cytosine and thymine bulges in duplex DNA.
  • Strongly and specifically binds to single cytosine and thymine bulges.
  • Stabilizes both single cytosine and thymine bulges in duplex DNA.
Catalog No. 50-003-39978 Supplier Medchemexpress LLC Supplier No. HY129594A10MG
DANP dihydrochloride is a bulge base recognition probe that strongly and specifically binds to the single cytosine and thymine bulges. It can stabilize both single cytosine and thymine bulges in duplex DNA.
  • Strongly and specifically binds to single cytosine and thymine bulges.
  • Stabilizes both single cytosine and thymine bulges in duplex DNA.