Learn More
Medchemexpress LLC DANP dihydrochloride | 99.3% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY129594A50MG
Encompass_Preferred
DANP dihydrochloride is a bulge base recognition probe that strongly and specifically binds to the single cytosine and thymine bulges. It can stabilize both single cytosine and thymine bulges in duplex DNA.
  • Bulge base recognition probe
  • Strongly and specifically binds to single cytosine and thymine bulges
  • Can stabilize both single cytosine and thymine bulges in duplex DNA
Catalog No. 50-003-41273