Learn More
Selleck Chemical LLC Danshensu S4741-20mg
SDP
Catalog No. 503108181
Encompass_Preferred
Danshensu (Salvianic acid A) a herbal preparation used in traditional Chinese medicine possesses potential antitumor and anti angiogenesis effects Danshensu inhibits CYP2E1 and CYP2C9 with IC50 of 36 63 and 75 76 m respectively
Catalog No. 50-310-8181 Supplier Selleck Chemical LLC Supplier No. S474120MG
Danshensu (Salvianic acid A) a herbal preparation used in traditional Chinese medicine possesses potential antitumor and anti angiogenesis effects Danshensu inhibits CYP2E1 and CYP2C9 with IC50 of 36 63 and 75 76 m respectively