Learn More
TARGETMOL CHEMICALS INC Danshensu sodium salt 100MG
SDP
Catalog No. 502876548
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Danshensu sodium salt (Sodium Danshensu) is sodium salt of danshensu from the widely used Chinese herb Danshen. It can inhibited phenylephrine- and CaCl2-induced vasoconstriction in Ca2+-free medium. Purity 97.04%
Catalog No. 50-287-6548 Supplier TARGETMOL CHEMICALS INC Supplier No. T2944100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Danshensu sodium salt (Sodium Danshensu) is sodium salt of danshensu from the widely used Chinese herb Danshen. It can inhibited phenylephrine- and CaCl2-induced vasoconstriction in Ca2+-free medium. Purity 97.04%