Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Danshenxinkun A | 65907-75-7 | MFCD08689944 | 5 MG
SDP

Catalog No. 5000418303
Encompass_Preferred

Danshenxinkun A is a natural compound isolated from Tanshen, utilized in studies related to heart diseases. It is intended for research use only.

  • Purity: 99.70%
  • Molecular weight: 296.32
  • Formula: C18H16O4
  • Appearance: Solid
  • Color: Light yellow to orange
  • Structure classification: Quinones, Naphthalene Quinones
  • Initial source: Plants
  • Solubility in DMSO: ≥ 100 mg/mL
  • In vivo solubility: 2.5 mg/mL for oral and intraperitoneal injection
  • Inhibits HIF1 activation in AGS cells (IC50 > 20 μM)
  • Inhibits HIF1 activation in Hep 3B2 cells (IC50 > 20 μM)
  • Does not significantly affect AGS cell viability under normoxic conditions (IC50 > 50 μM)
  • Does not significantly affect Hep 3B2 cell viability under normoxic conditions (IC50 > 50 μM)

Catalog No. 50-004-18303 Supplier Medchemexpress LLC Supplier No. HYN36805MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danshenxinkun A is a natural compound isolated from Tanshen, utilized in studies related to heart diseases. It is intended for research use only.

  • Purity: 99.70%
  • Molecular weight: 296.32
  • Formula: C18H16O4
  • Appearance: Solid
  • Color: Light yellow to orange
  • Structure classification: Quinones, Naphthalene Quinones
  • Initial source: Plants
  • Solubility in DMSO: ≥ 100 mg/mL
  • In vivo solubility: 2.5 mg/mL for oral and intraperitoneal injection
  • Inhibits HIF1 activation in AGS cells (IC50 > 20 μM)
  • Inhibits HIF1 activation in Hep 3B2 cells (IC50 > 20 μM)
  • Does not significantly affect AGS cell viability under normoxic conditions (IC50 > 50 μM)
  • Does not significantly affect Hep 3B2 cell viability under normoxic conditions (IC50 > 50 μM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.