Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dansyl Acid 98.0+%, TCI America™
SDP

Catalog No. D142625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D142625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D142625G Supplier TCI America Supplier No. D142625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4272-77-9
Molecular Formula C12H13NO3S
Molecular Weight (g/mol) 251.3
MDL Number MFCD00003995
InChI Key BBEQQKBWUHCIOU-UHFFFAOYSA-N
Synonym 5-Dimethylaminonaphthalene-1-sulfonic Acid
PubChem CID 1792
IUPAC Name 5-(dimethylamino)naphthalene-1-sulfonic acid
SMILES CN(C)C1=CC=CC2=C1C=CC=C2S(=O)(=O)O

Specifications

Melting Point 316°C
Color White-Yellow
Quantity 25 g
Formula Weight 251.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dansyl Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.