Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dansyl-Ala-Arg | 87687-46-5 | 99.36% | 478.57 | 1 MG
SDP

Catalog No. 5000397649
Encompass_Preferred

Dansyl-Ala-Arg is a synthetic peptide used for various biochemical studies.

  • Molecular weight: 478.57
  • Formula: C21H30N6O5S
  • Appearance: Off-white to light yellow solid
  • Sequence: {Dansyl}-Ala-Arg
  • Solubility: 50 mg/mL in DMSO

Catalog No. 50-003-97649 Supplier Medchemexpress LLC Supplier No. HYP43361MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dansyl-Ala-Arg is a synthetic peptide used for various biochemical studies.

  • Molecular weight: 478.57
  • Formula: C21H30N6O5S
  • Appearance: Off-white to light yellow solid
  • Sequence: {Dansyl}-Ala-Arg
  • Solubility: 50 mg/mL in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.