Learn More
Cayman Chemical Dansyl Amide, Purity: ≥98%, CAS: 1431-39-6, 1 g
SDP
Catalog No. 502912685
Encompass_Preferred
A fluorescent probe for albumin and CAII; binds to serum albumin and CAII (Kds = 7.57 and 0.2 µM, respectively); has been used to determine ligand affinities for these proteins; ex/em = 280/470 nm, respectively when bound to CAII
Catalog No. 50-291-2685 Supplier Cayman Chemical Supplier No. 361491 G
A fluorescent probe for albumin and CAII; binds to serum albumin and CAII (Kds = 7.57 and 0.2 µM, respectively); has been used to determine ligand affinities for these proteins; ex/em = 280/470 nm, respectively when bound to CAII