Learn More
Apexbio Technology LLC DANSYL-CHLORIDE-200MG
SDP
Catalog No. 5000569993
Encompass_Preferred
Dansyl chloride (CAS No 605-65-2) is a chemical reagent widely used in biochemical research particularly in fluorescence labeling applications Originating from sulfonamide derivatives it reacts with primary and secondary amines to form stable sulfonamide bonds enabling the visualization of biological molecules As a fluorescent probe it is highly effective in tracing biochemical substrates and pathways often operating in the nanomolar to low micromolar range Its applications include protein and peptide labeling aiding in the study of enzyme kinetics protein folding and cellular localization in various in vitro and in vivo models Dansyl chloride is also employed in drug screening processes to identify potential drug interactions and mechanisms Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-69993 Supplier Apexbio Technology LLC Supplier No. C6702200
Dansyl chloride (CAS No 605-65-2) is a chemical reagent widely used in biochemical research particularly in fluorescence labeling applications Originating from sulfonamide derivatives it reacts with primary and secondary amines to form stable sulfonamide bonds enabling the visualization of biological molecules As a fluorescent probe it is highly effective in tracing biochemical substrates and pathways often operating in the nanomolar to low micromolar range Its applications include protein and peptide labeling aiding in the study of enzyme kinetics protein folding and cellular localization in various in vitro and in vivo models Dansyl chloride is also employed in drug screening processes to identify potential drug interactions and mechanisms Experimental concentrations typically depend on the specific research design