Learn More
Aat Bioquest Dansyl chloride [5-Dimethylaminonaphthalene-1-sulfonyl chloride] *CAS 605-65-2*
SDP
Supplier:  Aat Bioquest 811
Encompass_Preferred
Dansyl chloride is non-fluorescent until it reacts with amines. The resulting Dansyl amides have environmentally sensitive fluorescence quantum yields and emission maxima along with large Stokes shifts.
Catalog No. 50-195-2948