Learn More
Medchemexpress LLC Dansyl chloride-d6 (DNSCl-d6) | 1276379-68-0 | 99.47% | 5 MG
SDP
Catalog No. 5000409700
Encompass_Preferred
Dansyl chloride-d6 is the deuterium labeled Dansyl chloride. It is a reagent that produces stable blue or blue-green fluorescent sulfonamide adducts in the reaction of aliphatic and aromatic amines with primary amino groups, and is widely used for modified amino acids, protein sequencing, and amino acid analysis. This compound is also utilized as a tracer and internal standard in various analytical methods.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-09700 Supplier Medchemexpress LLC Supplier No. HYD0017S5MG
Dansyl chloride-d6 is the deuterium labeled Dansyl chloride. It is a reagent that produces stable blue or blue-green fluorescent sulfonamide adducts in the reaction of aliphatic and aromatic amines with primary amino groups, and is widely used for modified amino acids, protein sequencing, and amino acid analysis. This compound is also utilized as a tracer and internal standard in various analytical methods.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.