Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dansyl Ethylenediamine, 5-Dimethylaminonaphthalene-1-(N-(2-Aminoethyl))sulfonamide

Catalog No. D112
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D112 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D112 Supplier Invitrogen™ Supplier No. D112
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

5-dimethylaminonaphthalene-1-(N-(2-aminoethyl))sulfonamide (dansyl ethylenediamine)

The primary aliphatic amine of dansyl ethylenediamine can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a generate stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3). Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247). This fluorophore is lipophilic and environmentally-sensitive.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mg
Product Type Dansyl Ethylenediamine
Content And Storage Store at room temperature and protect from light.
Chemical Reactivity Carboxylic Acid, Ketone, Aldehyde
Shipping Condition Room Temperature
Reactive Moiety Amine, Ethylenediamine
Label or Dye Dansyl
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.