Learn More
AdipoGen Dansyl fluoride
SDP
Catalog No. 502561169
Encompass_Preferred
Chemical. CAS 34523-28-9. Formula C12H12FNO2S. Molecular Weight 253.3. Dansyl fluoride is a fluorogenic reagent used as a specific fluorescent probe for serine proteases and is a serine-type protease inhibitor. It is an active site directed fluorescent inhibitor of guanidinobenzoatase GB, which binds to both the colonic isoenzymic forms of the GB. The GB on cell surfaces after binding dansyl fluoride makes the cells fluoresce blue. It has been used for the location of tumour cells in frozen sections of human lung squamous cell carcinoma and colonic carcinoma tissues. It is used as a precursor for the synthesis of other derivatives like dansyl amide. Spectral data lambdaex 360nm lambdaem 580nm methanol.
Catalog No. 50-256-1169 Supplier AdipoGen Supplier No. CDXD0241M250
Chemical. CAS 34523-28-9. Formula C12H12FNO2S. Molecular Weight 253.3. Dansyl fluoride is a fluorogenic reagent used as a specific fluorescent probe for serine proteases and is a serine-type protease inhibitor. It is an active site directed fluorescent inhibitor of guanidinobenzoatase GB, which binds to both the colonic isoenzymic forms of the GB. The GB on cell surfaces after binding dansyl fluoride makes the cells fluoresce blue. It has been used for the location of tumour cells in frozen sections of human lung squamous cell carcinoma and colonic carcinoma tissues. It is used as a precursor for the synthesis of other derivatives like dansyl amide. Spectral data lambdaex 360nm lambdaem 580nm methanol.