Learn More
Medchemexpress LLC HY-101131 5mg Medchemexpress, Dansyl glutathione CAS:75017-02-6 Purity:>98%
SDP
Catalog No. 502080617
Encompass_Preferred
Medchemexpress, HY-101131 5mg Dansyl glutathione CAS:75017-02-6 Dansyl glutathione is a trapping agent for the quantitative estimation and identification of reactive metabolites. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0617 Supplier Medchemexpress LLC Supplier No. HY1011315MG
Medchemexpress, HY-101131 5mg Dansyl glutathione CAS:75017-02-6 Dansyl glutathione is a trapping agent for the quantitative estimation and identification of reactive metabolites. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.