Learn More
Abcam Dansyl hydrazine, 100MG
Catalog No. 5051297
Encompass_Preferred
MW 265.33 Da, Purity >95%. Fluorescent lipophilic probe that reacts with aldehydes and ketones. Reacts with protein and water-soluble bipolymer carboxylic acids in aqueous solution using water-soluble carbodiimides. Used to label carbonyl groups prior to HPLC analysis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-512-97 Supplier Abcam Supplier No. AB274876100MG
Add to Cart
Add to Cart
MW 265.33 Da, Purity >95%. Fluorescent lipophilic probe that reacts with aldehydes and ketones. Reacts with protein and water-soluble bipolymer carboxylic acids in aqueous solution using water-soluble carbodiimides. Used to label carbonyl groups prior to HPLC analysis.
The product is subject to the following: Abcam Restricted Use Statement