Learn More
AdipoGen Dansyl hydrazine
SDP
Catalog No. 502561200
Encompass_Preferred
Chemical. CAS 33008-06-9. Formula C12H15N3O2S. MW 265.33. Dansyl hydrazine is a short-wavelength, UV-excited, fluorescent dye that has various applications in chemistry and biochemistry. It is a derivative of dansyl chloride, a compound commonly used for labeling and detecting amines and amino acids. Dansyl hydrazine itself is often employed for similar purposes. Dansyl hydrazine can be used to label primary and secondary amines in molecules, such as proteins and peptides. This labeling is useful for various applications, including protein analysis and drug development. Dansyl hydrazine is used to derivatize amino acids, making them more amenable to analysis by techniques like high-performance liquid chromatography HPLC and mass spectrometry MS. This allows for the quantification and identification of amino acids in complex biological samples. Dansyl hydrazine can react with carbonyl compounds, such as ketones and aldehydes, to form dansyl hydrazones.
Catalog No. 50-256-1200 Supplier AdipoGen Supplier No. CDXD0179G001
Chemical. CAS 33008-06-9. Formula C12H15N3O2S. MW 265.33. Dansyl hydrazine is a short-wavelength, UV-excited, fluorescent dye that has various applications in chemistry and biochemistry. It is a derivative of dansyl chloride, a compound commonly used for labeling and detecting amines and amino acids. Dansyl hydrazine itself is often employed for similar purposes. Dansyl hydrazine can be used to label primary and secondary amines in molecules, such as proteins and peptides. This labeling is useful for various applications, including protein analysis and drug development. Dansyl hydrazine is used to derivatize amino acids, making them more amenable to analysis by techniques like high-performance liquid chromatography HPLC and mass spectrometry MS. This allows for the quantification and identification of amino acids in complex biological samples. Dansyl hydrazine can react with carbonyl compounds, such as ketones and aldehydes, to form dansyl hydrazones.