Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dansyl Hydrazine 97.0+%, TCI America™
SDP

Catalog No. A55521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55521G 1 g
A55525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A55521G Supplier TCI America Supplier No. A55521G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 33008-06-9
Molecular Formula C12H15N3O2S
Molecular Weight (g/mol) 265.33
MDL Number MFCD00003986
InChI Key KPQYDVAFRDWIBW-UHFFFAOYSA-N
Synonym 5-Dimethylaminonaphthalene-1-sulfonyl Hydrazine
PubChem CID 94442
IUPAC Name 5-(dimethylamino)naphthalene-1-sulfonohydrazide
SMILES CN(C)C1=C2C=CC=C(C2=CC=C1)S(=O)(=O)NN

Specifications

Quantity 1 g
Formula Weight 265.33
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dansyl Hydrazine [for HPLC Labeling]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.