Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dansyl-L-isoleucine, TCI America™
SDP

Catalog No. D1497100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1497100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1497100MG Supplier TCI America Supplier No. D1497100MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 1100-21-6
Molecular Formula C18H24N2O4S
Molecular Weight (g/mol) 364.46
MDL Number MFCD00069525
InChI Key NJTOKQLVHXSRPR-UHFFFAOYNA-N
Synonym 5-Dimethylaminonaphthalene-1-sulfonyl-L-isoleucine, Dns-Ile-OH
PubChem CID 6602171
IUPAC Name 2-[5-(dimethylamino)naphthalene-1-sulfonamido]-3-methylpentanoic acid
SMILES CCC(C)C(NS(=O)(=O)C1=C2C=CC=C(N(C)C)C2=CC=C1)C(O)=O

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 364.46
Physical Form Crystalline Powder
Chemical Name or Material Dansyl-L-isoleucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.