Learn More
Medchemexpress LLC Dansyl-Tyr-Val-Gly | 113527-49-4 | 99.5% | 570.66 | 25 MG
SDP
Catalog No. 5000350707
Encompass_Preferred
Dansyl-Tyr-Val-Gly is a substrate of peptidylglycine monooxygenase (PHM) with a KD of 22 μM for PHM. It can be used to investigate the catalytic mechanism, kinetics, and inactivation characteristics of PHM. It also protects recombinant type A rat 75 kDa α-AE against ascorbate-mediated inactivation of its peptidyl-Gly hydroxylase activity and serves as a substrate for α-AE-mediated amidation to quantify residual enzyme activity.
  • Protects recombinant type A rat 75 kDa α-AE activity.
  • Serves as a substrate for α-AE-mediated amidation.
  • Powder stable for 2 years at -80°C and 1 year at -20°C.
  • Solution in solvent stable for 6 months at -80°C and 1 month at -20°C.
  • Store sealed, away from moisture.
Catalog No. 50-003-50707 Supplier Medchemexpress LLC Supplier No. 25MGHY138126
Dansyl-Tyr-Val-Gly is a substrate of peptidylglycine monooxygenase (PHM) with a KD of 22 μM for PHM. It can be used to investigate the catalytic mechanism, kinetics, and inactivation characteristics of PHM. It also protects recombinant type A rat 75 kDa α-AE against ascorbate-mediated inactivation of its peptidyl-Gly hydroxylase activity and serves as a substrate for α-AE-mediated amidation to quantify residual enzyme activity.
  • Protects recombinant type A rat 75 kDa α-AE activity.
  • Serves as a substrate for α-AE-mediated amidation.
  • Powder stable for 2 years at -80°C and 1 year at -20°C.
  • Solution in solvent stable for 6 months at -80°C and 1 month at -20°C.
  • Store sealed, away from moisture.