Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dansylamide 98.0+%, TCI America™
SDP

Catalog No. D540525MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D540525MG 25 mg
D5405100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D540525MG Supplier TCI America Supplier No. D540525MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1431-39-6
Molecular Formula C12H14N2O2S
Molecular Weight (g/mol) 250.316
MDL Number MFCD00004000
InChI Key TYNBFJJKZPTRKS-UHFFFAOYSA-N
Synonym dansylamide, dansyl amide, 5-dimethylamino naphthalene-1-sulfonamide, 5-dimethylamino-1-naphthalenesulfonamide, 1-naphthalenesulfonamide, 5-dimethylamino, 1okl, 5-dimethylamino naphthalene-1-sulphonamide, d0s7nh
PubChem CID 65077
IUPAC Name 5-(dimethylamino)naphthalene-1-sulfonamide
SMILES CN(C)C1=CC=CC2=C1C=CC=C2S(=O)(=O)N

Specifications

Melting Point 219°C
Color White-Yellow
Quantity 25 mg
Formula Weight 250.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dansylamide [for Albumin binding assay]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.