Learn More
Abcam 3-Dansylaminophenylboronic acid, 50MG
Catalog No. 50996105
Encompass_Preferred
MW 370.2 Da, Purity >98%. Fluorescent probe for cis-diols (sugars). Labels at high pH values on cell surfaces and in glycoproteins, which is used for in vitro carbohydrate determination and HPLC analysis of sugars. Attaches to vicinal diols of sugars, oligosaccharides and certain amino alcohols forming stable cyclic complexes. This agent is also an active site inhibitor for lipases and certain serine proteases. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-105 Supplier Abcam Supplier No. AB27431050MG
MW 370.2 Da, Purity >98%. Fluorescent probe for cis-diols (sugars). Labels at high pH values on cell surfaces and in glycoproteins, which is used for in vitro carbohydrate determination and HPLC analysis of sugars. Attaches to vicinal diols of sugars, oligosaccharides and certain amino alcohols forming stable cyclic complexes. This agent is also an active site inhibitor for lipases and certain serine proteases. .
The product is subject to the following: Abcam Restricted Use Statement