Learn More
Medchemexpress LLC HY-D1027 50mg Medchemexpress, Dansylcadaverine CAS:10121-91-2 Purity:>98%
SDP
Catalog No. 501933236
Encompass_Preferred
Medchemexpress, HY-D1027 50mg Dansylcadaverine CAS:10121-91-2 Dansylcadaverine is a fluorescent probe. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3236 Supplier Medchemexpress LLC Supplier No. HYD102750MG
Medchemexpress, HY-D1027 50mg Dansylcadaverine CAS:10121-91-2 Dansylcadaverine is a fluorescent probe. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.