Learn More
Sigma Aldrich Fine Chemicals Biosciences Dansylcadaverine 97 TLC1G
Catalog No. 502903978
Encompass
Dansylcadaverine also called as monodansylcadaverine (MDC) is a lysosomotropic agent. It is present in acidic and lipid rich regions. Dansylcadaverine also acts as a solvent polarity probe. It inhibits vesicular transport and decreases the rotavirus sequestering in L cells when pre-treated.
Catalog No. 50-290-3978 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D40081G
Add to Cart
Add to Cart
Dansylcadaverine also called as monodansylcadaverine (MDC) is a lysosomotropic agent. It is present in acidic and lipid rich regions. Dansylcadaverine also acts as a solvent polarity probe. It inhibits vesicular transport and decreases the rotavirus sequestering in L cells when pre-treated.