Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dansylglycine 98.0+%, TCI America™
SDP

Catalog No. D5406100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5406100MG 100 mg
D540625MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5406100MG Supplier TCI America Supplier No. D5406100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1091-85-6
Molecular Formula C14H16N2O4S
Molecular Weight (g/mol) 308.352
MDL Number MFCD00037734
InChI Key QHFXORCWAQTTGH-UHFFFAOYSA-N
Synonym N-[5-(Dimethylamino)naphthalene-1-sulfonyl]glycine, Dns-Gly-OH
PubChem CID 70666
IUPAC Name 2-[[5-(dimethylamino)naphthalen-1-yl]sulfonylamino]acetic acid
SMILES CN(C)C1=CC=CC2=C1C=CC=C2S(=O)(=O)NCC(=O)O

Specifications

Melting Point 159°C
Quantity 100 mg
Formula Weight 308.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dansylglycine [for Albumin binding assay]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.