Learn More
NATUS MEDICAL INC DANTEC DCN CONCENTRIC NEEDLE H
Catalog No. NC1832243
Encompass
Dantec DCN Concentric Needle Holder (Cable), 59" length.HUSH® shielded cable for Dantec DCN ElectrodesFits most EMG instrumentsLead Length: 59" (1.5 m)Electrode End: pogo pinInstrument End: 5-pin 240° DINPackaged: 1/pkg
This item is not returnable. View return policy
Catalog No. NC1832243 Supplier NATUS MEDICAL INC Supplier No. 9013C0014
Dantec DCN Concentric Needle Holder (Cable), 59" length.HUSH® shielded cable for Dantec DCN ElectrodesFits most EMG instrumentsLead Length: 59" (1.5 m)Electrode End: pogo pinInstrument End: 5-pin 240° DINPackaged: 1/pkg