Learn More
Apexbio Technology LLC DANTHRON-200MG
SDP
Catalog No. 5000569996
Encompass_Preferred
Danthron (CAS No 117-10-2) is a research compound often used in biomedical studies to explore its broad pharmacological effects Originating from anthraquinone derivatives Danthron functions primarily by influencing cellular pathways such as apoptosis and cell cycle arrest through the modulation of transcription factors and oxidative stress responses Its in vitro activity is typically observed in the nanomolar to low micromolar range reflecting its potency across various experimental settings Common applications include studies involving cancer cell lines where it is used to investigate antiproliferative effects and animal models to assess its potential therapeutic benefits Additionally Danthron is utilized in high-throughput drug screening assays due to its ability to target multiple cellular mechanisms Experimental concentrations typically depend on the specific research design allowing customization based on model systems and desired outcomes
Catalog No. 50-005-69996 Supplier Apexbio Technology LLC Supplier No. C6773200
Danthron (CAS No 117-10-2) is a research compound often used in biomedical studies to explore its broad pharmacological effects Originating from anthraquinone derivatives Danthron functions primarily by influencing cellular pathways such as apoptosis and cell cycle arrest through the modulation of transcription factors and oxidative stress responses Its in vitro activity is typically observed in the nanomolar to low micromolar range reflecting its potency across various experimental settings Common applications include studies involving cancer cell lines where it is used to investigate antiproliferative effects and animal models to assess its potential therapeutic benefits Additionally Danthron is utilized in high-throughput drug screening assays due to its ability to target multiple cellular mechanisms Experimental concentrations typically depend on the specific research design allowing customization based on model systems and desired outcomes