Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dantrolene-13C3 | 1185234-99-4 | 99.9% | 314.25 g/mol | C11(13C)3H10N4O5 | 1MG
SDP

Catalog No. 5000236122
Encompass_Preferred

Dantrolene-13C3 is a stable isotope-labeled analogue of dantrolene provided for research use as an internal standard or tracer in analytical and metabolic studies. The compound contains three 13C labels, has a molecular weight of 317.23 g/mol, and is typically supplied in milligram-scale quantities for analytical applications.

  • Stable isotope labeled with three 13C atoms.
  • High chemical purity (HPLC): 99.87%.
  • High isotopic enrichment: 99.1%.
  • Molecular weight: 317.23 g/mol.
  • CAS number: 1185234-99-4.
  • Supplied in milligram quantities suitable for analytical standards.

Catalog No. 50-002-36122 Supplier Medchemexpress LLC Supplier No. HY12542S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dantrolene-13C3 is a stable isotope-labeled analogue of dantrolene provided for research use as an internal standard or tracer in analytical and metabolic studies. The compound contains three 13C labels, has a molecular weight of 317.23 g/mol, and is typically supplied in milligram-scale quantities for analytical applications.

  • Stable isotope labeled with three 13C atoms.
  • High chemical purity (HPLC): 99.87%.
  • High isotopic enrichment: 99.1%.
  • Molecular weight: 317.23 g/mol.
  • CAS number: 1185234-99-4.
  • Supplied in milligram quantities suitable for analytical standards.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.