Learn More
TARGETMOL CHEMICALS INC DANTROLENE 50MG
SDP
Catalog No. 502579072
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dantrolene is a postsynaptic muscle relaxant by inhibiting Ca2+ ions release from sarcoplasmic reticulum stores by antagonizing ryanodine receptors. purity: 100%
Catalog No. 50-257-9072 Supplier TARGETMOL CHEMICALS INC Supplier No. T845250MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dantrolene is a postsynaptic muscle relaxant by inhibiting Ca2+ ions release from sarcoplasmic reticulum stores by antagonizing ryanodine receptors. purity: 100%