Learn More
Medchemexpress LLC Dantrolene | 7261-97-4 | MFCD00867709 | 98.4% | 314.25 g/mol | C14H10N4O5 | 10MM 1ML
SDP
Catalog No. 5000237874
Encompass_Preferred
Dantrolene is a ryanodine receptor antagonist and skeletal muscle relaxant offered as a ready-to-use 10 mM solution in DMSO (1 mL) and as solid pack sizes. The product is intended for use as a reference compound in in vitro pharmacology, biochemical assays, and RyR modulation studies; CAS 7261-97-4 and high assay purity support research applications.
  • Ready-to-use 10 mM solution in DMSO (1 mL) for in vitro assays.
  • Also available as solid in multiple pack sizes for formulation flexibility.
  • High purity (98.4%) confirmed by RP-HPLC.
  • Suitable as a ryanodine receptor antagonist and muscle relaxant reference.
  • Supplied with datasheet and HPLC purity report for quality assurance.
Catalog No. 50-002-37874 Supplier Medchemexpress LLC Supplier No. HY1254210MM 1ML
Dantrolene is a ryanodine receptor antagonist and skeletal muscle relaxant offered as a ready-to-use 10 mM solution in DMSO (1 mL) and as solid pack sizes. The product is intended for use as a reference compound in in vitro pharmacology, biochemical assays, and RyR modulation studies; CAS 7261-97-4 and high assay purity support research applications.
  • Ready-to-use 10 mM solution in DMSO (1 mL) for in vitro assays.
  • Also available as solid in multiple pack sizes for formulation flexibility.
  • High purity (98.4%) confirmed by RP-HPLC.
  • Suitable as a ryanodine receptor antagonist and muscle relaxant reference.
  • Supplied with datasheet and HPLC purity report for quality assurance.