Learn More
Medchemexpress LLC Dantrolene | 7261-97-4 | MFCD00867709 | 98.4% | 314.25 g/mol | C14H10N4O5 | 500MG
SDP
Catalog No. 5000235061
Encompass_Preferred
Dantrolene is a small-molecule ryanodine receptor antagonist used as a direct-acting skeletal muscle relaxant in research. It is supplied as a solid powder with a reported purity of 98.38% and is offered in multiple pack sizes, including a 500 mg vial. Manufacturer documentation includes a datasheet, certificate of analysis, and safety data sheet to support experimental use and handling.
Catalog No. 50-002-35061 Supplier Medchemexpress LLC Supplier No. HY12542500MG
Dantrolene is a small-molecule ryanodine receptor antagonist used as a direct-acting skeletal muscle relaxant in research. It is supplied as a solid powder with a reported purity of 98.38% and is offered in multiple pack sizes, including a 500 mg vial. Manufacturer documentation includes a datasheet, certificate of analysis, and safety data sheet to support experimental use and handling.