Learn More
Medchemexpress LLC Dantrolene | 7261-97-4 | MFCD00867709 | 98.4% | 314.26 | C14H10N4O5 | 5MG
SDP
Catalog No. 5000239629
Encompass_Preferred
Dantrolene is a research-grade skeletal muscle relaxant and ryanodine receptor (RyR) calcium channel modulator supplied as a high-purity solid or prepared solution for in vitro and analytical applications. The material is provided in small pack sizes suitable for biochemical assays, cellular studies, and method development; follow the supplier datasheet for handling, storage, and safety information.
  • High reported purity suitable for research assays.
  • Supplied in small quantities for low-usage experiments.
  • Available as solid or ready-to-use DMSO solution.
  • Suitable for biochemical and cellular RyR/calcium studies.
  • Includes datasheet and documentation for traceability.
  • Store according to supplier recommendations to maintain stability.
Catalog No. 50-002-39629 Supplier Medchemexpress LLC Supplier No. HY125425MG
Dantrolene is a research-grade skeletal muscle relaxant and ryanodine receptor (RyR) calcium channel modulator supplied as a high-purity solid or prepared solution for in vitro and analytical applications. The material is provided in small pack sizes suitable for biochemical assays, cellular studies, and method development; follow the supplier datasheet for handling, storage, and safety information.
  • High reported purity suitable for research assays.
  • Supplied in small quantities for low-usage experiments.
  • Available as solid or ready-to-use DMSO solution.
  • Suitable for biochemical and cellular RyR/calcium studies.
  • Includes datasheet and documentation for traceability.
  • Store according to supplier recommendations to maintain stability.