Learn More
Medchemexpress LLC Dantrolene | 7261-97-4 | MFCD00867709 | 98.4% | 314.26 g/mol | C14H10N4O5 | 10MG
SDP
Catalog No. 5000238148
Encompass_Preferred
Dantrolene is a research-grade skeletal muscle relaxant and ryanodine receptor (RyR) inhibitor used to study calcium homeostasis, malignant hyperthermia, muscle spasm, and related neurological or ischemic conditions. It acts as a non-competitive glutathione reductase inhibitor (reported Ki 111.6 μM; IC50 52.3 μM) and inhibits Ca2+ release from RyR1 and RyR3, making it useful for biochemical and cell-based studies.
  • Direct RyR1 and RyR3 calcium-release inhibitor.
  • Used in research on malignant hyperthermia and muscle spasm.
  • Non-competitive glutathione reductase inhibitor with reported Ki/IC50 values.
  • Available in solid and solution formats suitable for assays.
  • High reported purity (~98.4%).
  • Molecular formula C14H10N4O5; molecular weight ~314.26 g/mol.
Catalog No. 50-002-38148 Supplier Medchemexpress LLC Supplier No. HY1254210MG
Dantrolene is a research-grade skeletal muscle relaxant and ryanodine receptor (RyR) inhibitor used to study calcium homeostasis, malignant hyperthermia, muscle spasm, and related neurological or ischemic conditions. It acts as a non-competitive glutathione reductase inhibitor (reported Ki 111.6 μM; IC50 52.3 μM) and inhibits Ca2+ release from RyR1 and RyR3, making it useful for biochemical and cell-based studies.
  • Direct RyR1 and RyR3 calcium-release inhibitor.
  • Used in research on malignant hyperthermia and muscle spasm.
  • Non-competitive glutathione reductase inhibitor with reported Ki/IC50 values.
  • Available in solid and solution formats suitable for assays.
  • High reported purity (~98.4%).
  • Molecular formula C14H10N4O5; molecular weight ~314.26 g/mol.