Learn More
Selleck Chemical LLC Dantrolene sodium
SDP
Catalog No. 5000004018
Encompass_Preferred
Dantrolene sodium(F 440) acts as a postsynaptic muscle relaxant by inhibiting Ca2 ions release from sarcoplasmic reticulum stores by antagonizing ryanodine receptors
Catalog No. 50-000-04018 Supplier Selleck Chemical LLC Supplier No. S54781G
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Dantrolene sodium(F 440) acts as a postsynaptic muscle relaxant by inhibiting Ca2 ions release from sarcoplasmic reticulum stores by antagonizing ryanodine receptors