Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dantrolene (sodium hemiheptahydrate) | 24868-20-0 | MFCD07357270 | 99.8% | 399.31 g/mol | C14H16N4NaO8.5 | 10MM 1ML
SDP

Catalog No. 5000239630
Encompass_Preferred

Dantrolene sodium hemiheptahydrate is the sodium salt hydrate form of dantrolene used as a research reagent. It acts as a direct-acting skeletal muscle relaxant and inhibitor of calcium release from the sarcoplasmic reticulum, commonly used in studies of calcium homeostasis, malignant hyperthermia models, and muscle spasm pharmacology.

  • Purity 99.8% for reliable experimental consistency.
  • Molecular weight 399.31 g/mol and formula C14H16N4NaO8.5.
  • CAS number 24868-20-0 for substance traceability.
  • Solid, yellow to orange physical appearance.
  • Available in multiple pack sizes to suit laboratory needs.
  • Suitable for in vitro and in vivo research into calcium-related mechanisms.

Catalog No. 50-002-39630 Supplier Medchemexpress LLC Supplier No. HY12542A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dantrolene sodium hemiheptahydrate is the sodium salt hydrate form of dantrolene used as a research reagent. It acts as a direct-acting skeletal muscle relaxant and inhibitor of calcium release from the sarcoplasmic reticulum, commonly used in studies of calcium homeostasis, malignant hyperthermia models, and muscle spasm pharmacology.

  • Purity 99.8% for reliable experimental consistency.
  • Molecular weight 399.31 g/mol and formula C14H16N4NaO8.5.
  • CAS number 24868-20-0 for substance traceability.
  • Solid, yellow to orange physical appearance.
  • Available in multiple pack sizes to suit laboratory needs.
  • Suitable for in vitro and in vivo research into calcium-related mechanisms.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.