Learn More
Medchemexpress LLC Dantrolene sodium hemiheptahydrate | 24868-20-0 | MFCD07357270 | 99.8% | 399.31 g·mol⁻¹ | C14H9N4NaO5 · 3.5H2O | 500MG
SDP
Catalog No. 5000237876
Encompass_Preferred
Dantrolene sodium hemiheptahydrate is the sodium salt hydrate form of dantrolene, a direct-acting skeletal muscle relaxant and ryanodine receptor (RyR) inhibitor used as a research reagent in studies of muscle spasm, malignant hyperthermia, and calcium-related pathophysiology. Supplied in multiple pack sizes for laboratory use.
  • High purity (99.8%).
  • Inhibits RyR-mediated Ca2+ release for calcium signaling studies.
  • Supplied as the hemiheptahydrate crystalline form.
  • Molecular weight 399.31 g·mol⁻¹.
  • Available in multiple pack sizes, including 500 mg.
  • Intended for laboratory research use only.
Catalog No. 50-002-37876 Supplier Medchemexpress LLC Supplier No. HY12542A500MG
Dantrolene sodium hemiheptahydrate is the sodium salt hydrate form of dantrolene, a direct-acting skeletal muscle relaxant and ryanodine receptor (RyR) inhibitor used as a research reagent in studies of muscle spasm, malignant hyperthermia, and calcium-related pathophysiology. Supplied in multiple pack sizes for laboratory use.
  • High purity (99.8%).
  • Inhibits RyR-mediated Ca2+ release for calcium signaling studies.
  • Supplied as the hemiheptahydrate crystalline form.
  • Molecular weight 399.31 g·mol⁻¹.
  • Available in multiple pack sizes, including 500 mg.
  • Intended for laboratory research use only.