Learn More
Selleck Chemical LLC Danuglipron PF-06882961-5mg
SDP
Catalog No. 502389932
Encompass_Preferred
Danuglipron (PF-06882961), a non-peptide agonist, activates the canonical G protein signaling activity only in the Glucagon-like peptide-1 (GLP-1) receptor with Trp33ECD.
Catalog No. 50-238-9932 Supplier Selleck Chemical LLC Supplier No. S98515MG
Danuglipron (PF-06882961), a non-peptide agonist, activates the canonical G protein signaling activity only in the Glucagon-like peptide-1 (GLP-1) receptor with Trp33ECD.