Learn More
Apexbio Technology LLC Danusertib (PHA-739358), 5mg. Cas: 827318-97-8 MFCD: MFCD12024692. Pan-aurora kinase inhibitor
SDP
Supplier:  Apexbio Technology LLC A41165
Encompass_Preferred
Danusertib (previously known as PHA-739358), a 3-aminopyrazole derivative identified during the development of the pyrrolopyrazole sub-series, is a potent small-molecule inhibitor of aurora kinases family members with a dominant inhibition for aurora B kinase (ABK). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4885