Learn More
ATCC Daoy; Medulloblastoma; Human (Homo sapiens)
Catalog No. 502383557
Encompass
Daoy is a polygonal cell that was isolated from the cerebellum of a 4-year-old, White, male with desmoplastic cerebellar medulloblastoma. The biopsy tumor was taken from the posterior fossa. The cell line Daoy Med can be used in neuroscience research.
Catalog No. 50-238-3557 Supplier ATCC Supplier No. HTB186
Daoy is a polygonal cell that was isolated from the cerebellum of a 4-year-old, White, male with desmoplastic cerebellar medulloblastoma. The biopsy tumor was taken from the posterior fossa. The cell line Daoy Med can be used in neuroscience research.