Learn More
Aviva Systems Biology Corp DAP Antibody - middle region (ARP60156_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60156_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DAP. It was validated on Western Blot. . Alternate names: MGC99796ApplicationWB, IHC. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-286-964